missing translation for 'onlineSavingsMsg'
Learn More

Saflufenacil, TRC

Product Code. 30419632
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30419632 100 mg
30413067 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30419632 Supplier Toronto Research Chemicals Supplier No. TRCS081800100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372137-35-4
Molecular Formula C17 H17 Cl F4 N4 O5 S
Molecular Weight (g/mol) 500.85
Synonym Benzamide, 2-chloro-5-[3,6-dihydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)-1(2H)-pyrimidinyl]-4-fluoro-N-[[methyl(1-methylethyl)amino]sulfonyl]-, 2-Chloro-5-[3,6-dihydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)-1(2H)-pyrimidinyl]-4-fluoro-N-[[methyl(1-methylethyl)amino]sulfonyl]benzamide, 2-Chloro-4-fluoro-N-[isopropyl(methyl)sulfamoyl]-5-[3-methyl-2,6-dioxo-4-(trifluoromethyl)pyrimidin-1-yl]benzamide, 2-Chloro-5-[3,6-dihydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)-1(2H)-pyrimidinyl]-4-fluoro-N-[(isopropyl)methylsulfamoyl]benzamide, 2-Chloro-5-[3,6-dihydro-3-methyl-2,6-dioxo-4-trifluoromethyl-1(2H)-pyrimidinyl]-4-fluoro-N-[[N-methyl-N-(1-methylethyl)amino]sulfonyl]benzamide, Benmihuangcaoan, Eragon, Kixor, N'-[2-Chloro-4-fluoro-5-[1,2,3,6-tetrahydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)pyrimidin-1-yl]benzoyl]-N-isopropyl-N-methylsulfamide, Saflufenacil, Sharpen, Treevix
IUPAC Name 2-chloro-4-fluoro-5-[3-methyl-2,6-dioxo-4-(trifluoromethyl)pyrimidin-1-yl]-N-[methyl(propan-2-yl)sulfamoyl]benzamide
SMILES CC(C)N(C)S(=O)(=O)NC(=O)c1cc(N2C(=O)C=C(N(C)C2=O)C(F)(F)F)c(F)cc1Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H17ClF4N4O5S/c1-8(2)25(4)32(30,31)23-15(28)9-5-12(11(19)6-10(9)18)26-14(27)7-13(17(20,21)22)24(3)16(26)29/h5-8H,1-4H3,(H,23,28)
Formula Weight 500.0544
Chemical Name or Material Saflufenacil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.