Learn More
MEDICOM Safewear PP Gown 40 g/m2, Zip
Product Code. 31781374 Shop All Medicom Products
Change view
Click to view available options
Size:
Small
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781374 Small Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31781374 Supplier MEDICOM Supplier No. 40.635/02
  • ONLINE EXCLUSIVE PRICE
    60.64€
    49.72€ / Pack of 50
    Save 10.92€
    18% Off
Estimated Shipment: 09-12-2026
The polypropylene hygiene gown is comfortable and lightweight, with elasticated cuffs and zip closure to limit minor soiling risks.
  • Single-use hygiene coats protect the work environment from particles coming from the operator's body (scales, hair, sweat, etc/)
  • They are suitable for HACCP processes
  • They allow the operator to protect themselves from the risks of soiling related to the activity they carry out
Specifications
Size Small
Product Type Isolation Gown
Material Polypropylene
Color Blue
Closure Type Zipper
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic