Learn More
MEDICOM Safewear High Waterproof Overboots with Sole
Product Code. 31781346 Shop All Medicom Products
Change view
Click to view available options
Size:
Universal
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781346 Universal Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31781346 Supplier MEDICOM Supplier No. 10.204
  • ONLINE EXCLUSIVE PRICE
    162.24€
    133.01€ / Pack of 200
    Save 29.23€
    18% Off
Estimated Shipment: 07-12-2026
Low waterproof overshoes provide total protection against moisture and grip, even on wet floors.
  • Made of embossed PE, the overboot is 100% waterproof: tested according to the EN ISO 811 method for water penetration resistance under pressure
  • The sole ensures good grip on the ground, even on wet surfaces
  • Excellent abrasion resistance, even during prolonged use: tested according to the EN ISO 12947-2 method
  • Reinforced model in 200 μm embossed PE
  • Its foot-shaped (anatomical) design allows for easy fitting, even in uncomfortable positions
  • The overboot fully covers the shoe and calf of the user, fitting sizes from 34 to 48
  • Two tightening elastics ensure excellent support at the ankle and below the knee, preventing the overboot from slipping on the calf
Specifications
Color White
Material Polyethylene
Size Universal
Gender Unisex
Waterproof Yes
Product Type Overboot