Learn More
SCAT Safety Waste Cap 'LISA', V3.0 Extension Funnel
Product Code. 17346429 Shop All SCAT Europe Products
Change view
Click to view available options
Type:
Electrically Conductive
Not Electrically Conductive
Includes:
Extension Funnel
Extension Funnel with Earthing Cable
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Includes unitSize
17346429 Not Electrically Conductive Extension Funnel Each
17376429 Electrically Conductive Extension Funnel with Earthing Cable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17346429 Supplier SCAT Supplier No. 350120
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
Safety Waste Cap 'LISA', V3.0 Extension Funnel is an accessory for LISA HPLC/UHPLC systems.
  • Accessory for Safety Waste Cap 'LISA,' V3.0
  • Extension Funnel with Hinged Lid
  • Made of PTFE for optimal chemical resistance
Specifications
For Use With (Equipment) HPLC / UHPLC Systems
Includes Extension Funnel
Material PTFE
Thread Size M30x35
Type Not Electrically Conductive