Learn More
SCAT™ Safety Waste Cap 'LISA', V3.0
Shop All SCAT Europe Products
Click to view available options
Thread Size:
B53
B63
B83
G45
GL45
S50
S51
S55
S60/61
S65
S70
S70/71
S90
S95
Includes:
4 x 1.6/2.3/3.2 mm OD Fittings, 4 PFA Blind Plugs, 3 x 5 to 11.5 mm ID Fittings, 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
4 x 1.6/2.3/3.2 mm OD Fittings, 4 PFA Blind Plugs, 3 x 5 to 11.5 mm ID Fittings, 3 PTFE Blind Plugs
Unit Size:
1 set
Each
Description
The basic module includes a Safety Waste Cap V3.0 'LISA' with your selected thread size. Included in the scope of delivery are a further 23 accessories. In addition, a variety of other SCAT™ accessories such as a funnel or an exhaust air filter can be docked to the Safety Waste Cap LISA V3.0. You will find a selection listed in the ordering table:
- Satelite blind plug for extending and attaching various components such as a funnel
- Tube connector for connection for larger tubes with 5 to 11.5 mm ID
- Satellite made of PTFE for optimal chemical resistance
- 360° freely rotatable screw cap for easy changing of the container without twisting the tubing
- Fittings with improved design for easier connection of capillaries with 1.6 mm, 2.3 mm or 3.2 mm OD
- SCAT™ exhaust filters block harmful vapors with the help of optimized activated carbon and ensure safe pressure equalization in the waste containers
- Huge scope of delivery of fittings, blind plugs, and accessories ensures that docking system is ready for immediate use
Specifications
Specifications
| Includes | 4 x 1.6/2.3/3.2 mm OD Fittings, 4 PFA Blind Plugs, 3 x 5 to 11.5 mm ID Fittings, 3 PTFE Blind Plug, Blind Plug for Exhaust Filter |
| Material | PTFE |
| Thread Size | B53 |
| For Use With (Application) | HPLC and UHPLC Systems |
| Type | Waste Cap |