Order Info
dimSize
Specifications
Specifications
| Closure Size | S60/61 |
| Description | Flexible |
| Color | Translucent White |
| For Use With (Application) | Simple and Safe Pouring of Fluids From Containers |
| For Use With (Equipment) | Canister, Container & Bottles |
| Material | HDPE |
| Product Type | Safety Spout |