missing translation for 'onlineSavingsMsg'
Learn More

Ekastu Safety™ M3-PLUS Chemical Protection Gloves

Product Code. 10478982
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
10478982 9 Each
10141811 7 Each
10234631 8 Each
10623822 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10478982 Supplier Ekastu Safety™ Supplier No. 481122

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Resistant to many chemical substances such as oils and fats, pesticides, solvents and fuels as well as bacteriological contamination. EKASTU Safety™ M3-PLUS Chemical Protection Gloves are made of nitrile perbunan (NBR).

  • Category 3 Classification
  • Color: green
  • Made of nitrile perbunan (NBR)
  • Anatomical shape with very good gropeability
  • Sanitized, cotton lined, patterned for a non-slip grip
  • Cotton flocked
  • Non-abrasive
  • Resistant to many chemical substances such as oils and fats, pesticide, solvents and fuels as well as bacteriological contamination
  • Maximum abrasion resistance according to DIN EN 388, thereby maximum service life

Specifications

Certifications/Compliance DIN EN 388
Classification Category 3
Color Green
Material Nitrile
Powdered Powder Free
Product Type Gloves
Size 9
Sterility Non-sterile
Finger Style Textured
Glove Cuff Style Open
Lining Cotton-flocked
Palm Type Textured
Quantity 1 Each
Resistant To Abrasion Resistant
Length (Metric) 31 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.