Learn More
Ekastu Safety™ M3-PLUS Chemical Protection Gloves
Description
- Category 3 Classification
- Color: green
- Made of nitrile perbunan (NBR)
- Anatomical shape with very good gropeability
- Sanitized, cotton lined, patterned for a non-slip grip
- Cotton flocked
- Non-abrasive
- Resistant to many chemical substances such as oils and fats, pesticide, solvents and fuels as well as bacteriological contamination
- Maximum abrasion resistance according to DIN EN 388, thereby maximum service life
Specifications
Specifications
| Certifications/Compliance | DIN EN 388 |
| Classification | Category 3 |
| Color | Green |
| Material | Nitrile |
| Powdered | Powder Free |
| Product Type | Gloves |
| Size | 7 |
| Sterility | Non-sterile |
| Finger Style | Textured |
| Glove Cuff Style | Open |
| Show More |