missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Specifications
Antigen | S100A4 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16815604
|
Proteintech
16105-1-AP-20UL |
20 μL |
140.00€
20µL |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
16805604
|
Proteintech
16105-1-AP-150UL |
150 μL |
399.00€
150µL |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
Description
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.Specifications
S100A4 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P07091, P26447 | |
20198, 6275 | |
S100A4 Fusion Protein Ag9019 | |
Primary | |
-20°C | |
S100A4 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
18A2, 42A, Calvasculin, CAPL, Fibroblast Specific Protein, Fibroblast-specific protein 1, FSP1, FSP-1, Metastasin, MTS1, P9KA, PEL98, Protein Mts1, Protein S100 A4, S100A4 | |
S100A4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |