Learn More
(S)-tert-Butyl 2-amino-4-methylpentanoate hydrochloride, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 2748-02-9 |
| Molecular Formula | C10H22ClNO2 |
| MDL Number | MFCD00038906 |
| Quantity | 500 g |
| SMILES | CC(C)C[C@H](N)C(OC(C)(C)C)=O.Cl |
| Molecular Weight (g/mol) | 223.74 |
| Percent Purity | 98% |
| Chemical Name or Material | (S)-tert-Butyl 2-amino-4-methylpentanoate hydrochloride |