Learn More
(S)-SPINOL, TRC
Specifications
Specifications
| Chemical Name or Material | 3,3'-spirobi[1,2-dihydroindene]-4,4'-diol |
| CAS | 223259-63-0 |
| Molecular Formula | C17 H16 O2 |
| Quantity | 1 g |
| Synonym | (S)-SPINOL, (1S)-2,2',3,3'-Tetrahydro-1,1'-spirobi[1H-indene]-7,7'-diol, (S)-1,1'-Spirobiindane-7,7'-diol |
| InChI Formula | InChI=1S/C17H16O2/c18-13-5-1-3-11-7-9-17(15(11)13)10-8-12-4-2-6-14(19)16(12)17/h1-6,18-19H,7-10H2 |
| SMILES | Oc1cccc2CCC3(CCc4cccc(O)c34)c12 |
| IUPAC Name | 3,3'-spirobi[1,2-dihydroindene]-4,4'-diol |
| Molecular Weight (g/mol) | 252.308 |
| Formula Weight | 252.115 |