Learn More
(S,R)-Palonosetron Hydrochloride Contains up to ~35% (R,R)-isomer, TRC
Product Code. 30136167
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136167 50 mg 50mg
30004005 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30136167 Supplier Toronto Research Chemicals Supplier No. TRCP16581050MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 135729-76-9
Molecular Formula C19 H24 N2 O . Cl H
Molecular Weight (g/mol) 332.87
Synonym 1H-Benz[de]isoquinolin-1-one, 2-(3R)-1-azabicyclo[2.2.2]oct-3-yl-2,3,3a,4,5,6-hexahydro-, hydrochloride (1:1), (3aS)-, 1H-Benz[de]isoquinolin-1-one, 2-(1-azabicyclo[2.2.2]oct-3-yl)-2,3,3a,4,5,6-hexahydro-, monohydrochloride, [R-(R*,S*)]- (9CI), (3aS)-2-(3R)-1-Azabicyclo[2.2.2]oct-3-yl-2,3,3a,4,5,6-hexahydro-1H-benz[de]isoquinolin-1-one hydrochloride (1:1), RS 25233-197
IUPAC Name (3aS)-2-[(3R)-1-azabicyclo[2.2.2]octan-3-yl]-3a,4,5,6-tetrahydro-3H-benzo[de]isoquinolin-1-one;hydrochloride
SMILES Cl.O=C1N(C[C@H]2CCCc3cccc1c23)[C@H]4CN5CCC4CC5
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C19H24N2O.ClH/c22-19-16-6-2-4-14-3-1-5-15(18(14)16)11-21(19)17-12-20-9-7-13(17)8-10-20;/h2,4,6,13,15,17H,1,3,5,7-12H2;1H/t15-,17+;/m1./s1
Formula Weight 332.1655
Chemical Name or Material (S,R)-Palonosetron Hydrochloride