Learn More
(S)-(+)-Pregabalin-d4 (aminomethyl-d2; hexanoic-2,2-d2 acid), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Pregabalin-d4 |
| CAS | 1276197-54-6 |
| Molecular Formula | C8 D4 H13 N O2 |
| Quantity | 0.01 g |
| Synonym | (3 S)-3-(Aminomethyl-d2)-5-methylhexanoic-2,2-d2 acid, Pregabalin-d4, Pregabalin-D4 (4-aminobutanoic-2,2,4,4-D4), (S)-(+)-Pregabalin-d4 (aminomethyl-d2, hexanoic-2,2-d2 acid), (3 S)-3-[Amino(dideuterio)methyl]-2,2-dideuterio-5-methylhexanoic acid |
| InChI Formula | InChI=1 S/C8H17NO2/c1-6(2)3-7(5-9)4-8(10)11/h6-7 H,3-5,9H2,1-2H3,(H,10,11)/t7-/m0/s1/i4D2,5D2 |
| SMILES | [2 H]C([2 H])(N)[C@@H](CC(C)C)C([2 H])([2 H])C(=O)O |
| IUPAC Name | (3 S)-3-[amino(dideuterio)methyl]-2,2-dideuterio-5-methylhexanoic acid |
| Molecular Weight (g/mol) | 163.2507 |
| Formula Weight | 163.151 g/mol |
| Show More |