missing translation for 'onlineSavingsMsg'
Learn More

(S)-Pramipexole, TRC

Product Code. 30428470
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30428470 2.5 g
30416926 10 g
30419999 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428470 Supplier Toronto Research Chemicals Supplier No. TRCP7007502.5G
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104632-26-0
Molecular Formula C10H17N3S
Molecular Weight (g/mol) 211.33
Synonym 2,6-Benzothiazolediamine, 4,5,6,7-tetrahydro-N6-propyl-, (6S)-, 2,6-Benzothiazolediamine, 4,5,6,7-tetrahydro-N6-propyl-, (S)-, (-)-Pramipexole, (S)-2-Amino-6-propylamino-4,5,6,7-tetrahydrobenzothiazole, (S)-Pramipexole, Mirapexin, Pramipexole, SND 919, SUD 919CL2Y, Sifrol, U 98528E
IUPAC Name (6S)-6-N-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine
SMILES CCCN[C@H]1CCc2nc(N)sc2C1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C10H17N3S/c1-2-5-12-7-3-4-8-9(6-7)14-10(11)13-8/h7,12H,2-6H2,1H3,(H2,11,13)/t7-/m0/s1
Formula Weight 211.1143
Chemical Name or Material Pramipexole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.