Learn More
S-Phos, TRC
Product Code. 30412908
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412908 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30412908 Supplier Toronto Research Chemicals Supplier No. TRCS681720500MG
 Restricted Item
Estimated Shipment: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material S-Phos
CAS 657408-07-6
Molecular Formula C26H35O2P
Quantity 500 mg
Synonym (2,6-Dimethoxy-1,1'-biphenyl-2-yl)dicyclohexylphosphine, 2-(Dicyclohexylphosphino)-2',6'-dimethoxy-1,1'-biphenyl, 2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl, Dicyclohexyl(2',6'-dimethoxy-1,1'-biphenyl-2-yl)phosphine, Dicyclohexyl(2',6'-dimethoxybiphenyl-2-yl)phosphine
InChI Formula InChI=1S/C26H35O2P/c1-27-23-17-11-18-24(28-2)26(23)22-16-9-10-19-25(22)29(20-12-5-3-6-13-20)21-14-7-4-8-15-21/h9-11,16-21H,3-8,12-15H2,1-2H3
SMILES COc1cccc(OC)c1c2ccccc2P(C3CCCCC3)C4CCCCC4
IUPAC Name dicyclohexyl-[2-(2,6-dimethoxyphenyl)phenyl]phosphane
Molecular Weight (g/mol) 410.53
Formula Weight 410.2375