missing translation for 'onlineSavingsMsg'
Learn More

(S)-Nornicotine, TRC

Product Code. 30426271
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426271 100 mg 100mg
30422354 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30426271 Supplier Toronto Research Chemicals Supplier No. TRCN756995100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-Nornicotine
CAS 494-97-3
Purity Grade Notes HPLC
Molecular Formula *C3C6H12N2
Quantity 100 mg
Synonym Pyridine','3-(2S)-2-pyrrolidinyl-, Nicotine','1'-demethyl- (8CI), Pyridine','3-(2-pyrrolidinyl)-','(S)-, 3-(2S)-2-Pyrrolidinylpyridine, (-)-(S)-Nornicotine, (-)-Nornicotine, (S)-(-)-Nornicotine, (S)-2-(3-Pyridyl)pyrrolidine, (S)-Nornicotine, 1'-Demethylnicotine, Nornicotin, Nornicotine, l-Nornicotine
InChI Formula InChI=1S/C9H12N2/c1-3-8(7-10-5-1)9-4-2-6-11-9/h1,3,5,7,9,11H,2,4,6H2/t9-/m0/s1
SMILES C1CN[C@@H](C1)c2cccnc2
IUPAC Name 3-[(2S)-pyrrolidin-2-yl]pyridine
Molecular Weight (g/mol) 151.18
Formula Weight 148.1
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.