Learn More
S-Nitroso-L-glutathione (>90%), TRC
Product Code. 30135625
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135625 50 mg 50mg
30135551 25 mg 25mg
30007092 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135625 Supplier Toronto Research Chemicals Supplier No. TRCN52750050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 57564-91-7
Synonym (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-3-nitrososulfanyl-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
Specifications
Format Neat
Molecular Formula C10H16N4O7S
Molecular Weight (g/mol) 336.32
InChI Key HYHSBSXUHZOYLX-WDSKDSINSA-N
SMILES N[C@@H](CCC(=O)N[C@@H](CSN=O)C(=O)NCC(O)=O)C(O)=O
IUPAC Name (2S)-2-amino-4-{[(1R)-1-[(carboxymethyl)carbamoyl]-2-(nitrososulfanyl)ethyl]carbamoyl}butanoic acid
Quantity 50 mg
References Thompson, C., et al.: Toxicol., App. Pharmacol., 233, 355 (2008),
Shipping Condition Room Temperature