Learn More
(S)-N-[2-(5-Bromo-2,3-dihydro-6-methoxy-1H-inden-1-yl)ethyl]propanamide, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-N-[2-(5-Bromo-2,3-dihydro-6-methoxy-1H-inden-1-yl)ethyl]propanamide |
| CAS | 196597-83-8 |
| Molecular Formula | C15H20BrNO2 |
| Quantity | 25 mg |
| Synonym | N-[2-[(1S)-5-Bromo-2,3-dihydro-6-methoxy-1H-inden-1-yl]ethyl]propanamide |
| InChI Formula | InChI=1S/C15H20BrNO2/c1-3-15(18)17-7-6-10-4-5-11-8-13(16)14(19-2)9-12(10)11/h8-10H,3-7H2,1-2H3,(H,17,18)/t10-/m0/s1 |
| SMILES | CCC(=NCC[C@@H]1CCc2cc(Br)c(OC)cc12)O |
| IUPAC Name | N-[2-[(1S)-5-bromo-6-methoxy-2,3-dihydro-1H-inden-1-yl]ethyl]propanamide |
| Molecular Weight (g/mol) | 326.23 |
| Formula Weight | 325.0677 |