Learn More
(S)-N-(1-Amino-5-guanidino-1-oxopentan-2-yl)benzamide hydrochloride, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 4299-03-0 |
| Molecular Formula | C13H20ClN5O2 |
| MDL Number | MFCD00035003 |
| Quantity | 5 g |
| SMILES | O=C(N[C@@H](CCCNC(N)=N)C(N)=O)C1=CC=CC=C1.[H]Cl |
| Molecular Weight (g/mol) | 313.78 |
| Percent Purity | 95% |
| Chemical Name or Material | (S)-N-(1-Amino-5-guanidino-1-oxopentan-2-yl)benzamide hydrochloride |