Learn More
S-Methyl-D-penicillamine, TRC
Specifications
Specifications
| Chemical Name or Material | S-Methyl-D-penicillamine |
| CAS | 29913-84-6 |
| Molecular Formula | C6H13NO2S |
| Quantity | 1 g |
| Synonym | 3-(Methylthio)-D-valine, S-Methylpenicillamine |
| InChI Formula | InChI=1S/C6H13NO2S/c1-6(2,10-3)4(7)5(8)9/h4H,7H2,1-3H3,(H,8,9)/t4-/m0/s1 |
| SMILES | CC(C)([C@H](C(=O)O)N)SC |
| IUPAC Name | (2S)-2-amino-3-methyl-3-methylsulfanylbutanoic acid |
| Molecular Weight (g/mol) | 163.24 |
| Formula Weight | 163.0667 |