missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (S)-MCPG
GREENER_CHOICE

Product Code. p-4783140
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001081

Brand: Tocris Bioscience™ 0337/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Non-selective mGlu antagonist; active isomer of (RS)-MCPG (Cat. No. 0336)

Active isomer of (RS)-MCPG . Non-selective group I/group II metabotropic glutamate receptor antagonist.

Available as part of the Group I mGlu Receptor Tocriset™, Group II mGlu Receptor Tocriset™ and Mixed mGlu Receptor Tocriset™ . Racemate also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150145-89-4
Molecular Formula C10H10NO4

Specifications

Quantity 10 mg
Formula Weight 209.2
Purity >99%
Molecular Weight (g/mol) 208.19
Chemical Name or Material (S)-MCPG
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.