Learn More
Heidolph Instruments S-Box Package 1
Product Code. 17868000 Shop All Heidolph Instruments Products
Click to view available options
Color:
Blue
Green
Orange
Violet
Unit Size:
Each
Product Code. 17868000
Brand: Heidolph Instruments 4000500200
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The S-Box Package 1 includes additional accessories to complete an existing S-Biosystem incubation system.
  • Each S-box is a stand-alone miniature incubator.
  • Inside the S-box there is room for one standard multi-well plate, or one cell culture bottle (up to T75).
  • The complete interior (inner box) can be purchased as a sterilized exchangeable element in a sterile outer packaging.
  • Your culture is prevented from drying out by a sachet of sterilized water (Waterbag), which is enough for weeks.
  • To avoid mix-up, the front cover of the S-box is available in different colors.
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Includes 1 S-box, 1 Tool Kit, 4 inner boxes, 4 water bags, 4 front covers