Learn More
(S)-Bethanechol, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-Bethanechol |
| CAS | 944538-50-5 |
| Molecular Formula | C7H17ClN2O2 |
| Quantity | 10 mg |
| Synonym | (2S)-2-[(Aminocarbonyl)oxy]-N,N,N-trimethyl-1propanaminium Chloride |
| InChI Formula | InChI=1S/C7H16N2O2.ClH/c1-6(11-7(8)10)5-9(2,3)4;/h6H,5H2,1-4H3,(H-,8,10);1H/t6-;/m0./s1 |
| SMILES | [Cl-].C[C@@H](C[N+](C)(C)C)OC(=O)N |
| IUPAC Name | [(2S)-2-carbamoyloxypropyl]-trimethylazanium;chloride |
| Molecular Weight (g/mol) | 196.68 |
| Formula Weight | 196.0979 |