missing translation for 'onlineSavingsMsg'
Learn More

(S)-Apomorphine Hydrochloride, TRC

Product Code. 18070200
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18070200 10 mg 10mg
18051720 5 mg 5mg
17989829 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18070200 Supplier Toronto Research Chemicals Supplier No. TRCA72750510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41035-30-7
Molecular Formula C17 H17 N O2 . H Cl
Molecular Weight (g/mol) 303.78
Synonym 4H-Dibenzo[de,g]quinoline-10,11-diol, 5,6,6a,7-tetrahydro-6-methyl-, hydrochloride (1:1), (6aS)-, 4H-Dibenzo[de,g]quinoline-10,11-diol, 5,6,6a,7-tetrahydro-6-methyl-, hydrochloride, (S)- (9CI), (6aS)-10,11-Dihydroxyaporphine hydrochloride, (S)-(+)-Apomorphine hydrochloride, (S)-Apomorphine hydrochloride, (S)-6-Methyl-5,6,6a,7-tetrahydro-4H-dibenzo[de,g]quinoline-10,11-diol hydrochloride, (S)-5,6,6a,7-Tetrahydro-6-methyl- 4H-dibenzo[de,g]quinoline-10,11-diol Hydrochloride;
IUPAC Name (6aS)-6-methyl-5,6,6a,7-tetrahydro-4H-dibenzo[de,g]quinoline-10,11-diol;hydrochloride
SMILES Cl.CN1CCc2cccc3c2[C@@H]1Cc4ccc(O)c(O)c34

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H17NO2.ClH/c1-18-8-7-10-3-2-4-12-15(10)13(18)9-11-5-6-14(19)17(20)16(11)12;/h2-6,13,19-20H,7-9H2,1H3;1H/t13-;/m0./s1
Formula Weight 303.1026
Chemical Name or Material (S)-Apomorphine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.