missing translation for 'onlineSavingsMsg'
Learn More

S-Acetylthiocholine iodide, 98%

Product Code. 11472828
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11472828

Brand: Thermo Scientific Alfa Aesar A16802.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1866-15-5 | C7H16INOS | 289.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1866-15-5
Molecular Formula C7H16INOS
Molecular Weight (g/mol) 289.175
MDL Number MFCD00011819
InChI Key NTBLZMAMTZXLBP-UHFFFAOYSA-M
Synonym acetylthiocholine iodide, s-acetylthiocholine iodide, acetylthiocholine diiodide, 2-acetylthioethyltrimethylammonium iodide, 2-acetylthio-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-acetylthio-n,n,n-trimethyl-, iodide, acetylthiocholinjodid, choline, s-acetylthio-, iodide, 2-acetylsulfanyl-n,n,n-trimethylethanaminium iodide, acetyl thiobilineurine iodide
PubChem CID 74629
IUPAC Name 2-acetylsulfanylethyl(trimethyl)azanium;iodide
SMILES CC(=O)SCC[N+](C)(C)C.[I-]

Specifications

Melting Point 205°C to 209°C
Quantity 5 g
UN Number UN2811
Beilstein 3916578
Sensitivity Hygroscopic; Light sensitive
Formula Weight 289.18
Percent Purity 98%
Chemical Name or Material S-Acetylthiocholine iodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.