Learn More
S-Acetyl-L-glutathione, TRC
Specifications
Specifications
| Chemical Name or Material | S-Acetylglutathione |
| CAS | 3054-47-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12 H19 N3 O7 S |
| Quantity | 1 g |
| Synonym | S-Acetyl-L-glutathione, L-γ-glutamyl-S-acetyl-L-cysteinyl-Glycine, Glutathione','S-acetate (6CI,7CI), Glutathione','acetate (ester) (8CI), Glycine','N-(S-acetyl-N-L-γ-glutamyl-L-cysteinyl)-, S-Acetylglutathione |
| InChI Formula | InChI=1S/C12H19N3O7S/c1-6(16)23-5-8(11(20)14-4-10(18)19)15-9(17)3-2-7(13)12(21)22/h7-8H,2-5,13H2,1H3,(H,14,20)(H,15,17)(H,18,19)(H,21,22)/t7-,8-/m0/s1 |
| SMILES | CC(=O)SC[C@H](NC(=O)CC[C@H](N)C(=O)O)C(=O)NCC(=O)O |
| IUPAC Name | (2S)-5-[[(2R)-3-acetylsulfanyl-1-(carboxymethylamino)-1-oxopropan-2-yl]amino]-2-amino-5-oxopentanoic acid |
| Molecular Weight (g/mol) | 349.36 |
| Show More |