Learn More
(S)-4-Hydroxy-4-methyl-7,8-dihydro-1H-pyrano[3,4-f]-indolizine-3,6,10(4H)-trione, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-4-Hydroxy-4-methyl-7,8-dihydro-1H-pyrano[3,4-f]-indolizine-3,6,10(4H)-trione |
| CAS | 1332605-57-8 |
| Molecular Formula | C9 H8 O7 S . 2 Na |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C9H10O7S.2Na/c1-15-8-4-6(5-9(10)11)2-3-7(8)16-17(12,13)14;;/h2-4H,5H2,1H3,(H,10,11)(H,12,13,14);;/q;2*+1/p-2 |
| SMILES | [Na+].[Na+].COc1cc(CC(=O)[O-])ccc1OS(=O)(=O)[O-] |
| IUPAC Name | disodium;2-(3-methoxy-4-sulfonatooxyphenyl)acetate |
| Molecular Weight (g/mol) | 306.2 |
| Formula Weight | 305.979 |