missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (S)-4-Carboxyphenylglycine
GREENER_CHOICE

Product Code. p-4783137
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001077

Brand: Tocris Bioscience™ 0323/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Competitive group I mGlu antagonist; also weak group II agonist

(S)-4-Carboxyphenylglycine is a competitive group I metabotropic glutamate receptor antagonist, with selectivity for mGlu1a/1a over mGlu5a/5b.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134052-73-6
Molecular Formula C9H9NO4

Specifications

Quantity 10 mg
Formula Weight 195.17
Purity >99%
Molecular Weight (g/mol) 195.174
Chemical Name or Material (S)-4-Carboxyphenylglycine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.