Learn More
(S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-1-naphthalenol (Mixture of Diastereomers), TRC
Specifications
Specifications
| Chemical Name or Material | (S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-1-naphthalenol (Mixture of Diastereomers) |
| CAS | 374777-87-4 |
| Molecular Formula | C16 H14 Cl2 O |
| Quantity | 100 mg |
| Synonym | Sertraline Impurity 16 |
| InChI Formula | InChI=1S/C16H14Cl2O/c17-14-7-5-10(9-15(14)18)11-6-8-16(19)13-4-2-1-3-12(11)13/h1-5,7,9,11,16,19H,6,8H2/t11-,16?/m0/s1 |
| SMILES | OC1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13 |
| IUPAC Name | (4S)-4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydronaphthalen-1-ol |
| Molecular Weight (g/mol) | 293.188 |
| Formula Weight | 292.042 |