Learn More
S-(3-Carboxypropyl)-L-cysteine, TRC
Product Code. 30360421
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30360421 100 mg
30350966 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30360421 Supplier Toronto Research Chemicals Supplier No. TRCC181360100MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 30845-11-5
Molecular Formula C7H13NO4S
Molecular Weight (g/mol) 207.25
Synonym (R)-4-[(2-Amino-2-carboxyethyl)thio]butanoic Acid, L-4-[(2-Amino-2-carboxyethyl)thio]butyric Acid, 3-(3-Carboxypropylthio)alanine
IUPAC Name 4-[(2R)-2-amino-2-carboxyethyl]sulfanylbutanoic acid
SMILES C(CC(=O)O)CSC[C@@H](C(=O)O)N
Specifications
InChI Formula InChI=1S/C7H13NO4S/c8-5(7(11)12)4-13-3-1-2-6(9)10/h5H,1-4,8H2,(H,9,10)(H,11,12)/t5-/m0/s1
Quantity 100 mg
Formula Weight 207.0565
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material S-(3-Carboxypropyl)-L-cysteine
Grade HPLC