Learn More
(S)-3-Amino-6,7-dihydroxy-hydrocoumarin Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-3-Amino-6,7-dihydroxy-hydrocoumarin Hydrochloride |
| CAS | 30033-29-5 |
| Molecular Formula | C9H10ClNO4 |
| Quantity | 100 mg |
| Synonym | (S)-3-Amino-6,7-dihydroxy-hydrocoumarin Hydrochloride, (S)-3-Amino-3,4-dihydro-6,7-dihydroxy-2H-1-Benzopyran-2-one Hydrochloride |
| InChI Formula | InChI=1S/C9H9NO4.ClH/c10-5-1-4-2-6(11)7(12)3-8(4)14-9(5)13;/h2-3,5,11-12H,1,10H2;1H/t5-;/m0./s1 |
| SMILES | O=C1OC2=CC(O)=C(O)C=C2C[C@@H]1N.Cl |
| IUPAC Name | (S)-3-amino-6,7-dihydroxychroman-2-one hydrogen chloride |
| Molecular Weight (g/mol) | 231.63 |
| Formula Weight | 231.03 |