missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-4-(2,4,5-trifluorophenyl)butanoic Acid Hydrochloride, TRC

Product Code. 30322382
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322382 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322382 Supplier Toronto Research Chemicals Supplier No. TRCA168940100MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-3-Amino-4-(2,4,5-trifluorophenyl)butanoic Acid Hydrochloride
CAS 1217809-78-3
Molecular Formula C10H10F3NO2 . HCl
Quantity 100 mg
Synonym (S)-3-Amino-4-(2,4,5-trifluorophenyl)butanoic Acid Hydrochloride, (S)-3-Amino-4-(2,4,5-trifluorophenyl)-butyric Acid-HCl
InChI Formula InChi=S/C10H10F3NO2.ClH/c11-7-4-9(13)8(12)2-5(7)1-6(14)3-10(15)16;/h2,4,6H,1,3,14H2,(H,15,16);1H/t6-;/m0./s1
SMILES FC1=CC(C[C@H](N)CC(O)=O)=C(F)C=C1F.Cl
IUPAC Name 3S)-3-amino-4-(2,4,5-trifluorophenyl)butanoic acid hydrochloride
Molecular Weight (g/mol) 269.65
Formula Weight 269.05
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.