missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (S)-3,5-DHPG
GREENER_CHOICE

Product Code. p-4783133
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 18745936

Brand: Tocris Bioscience™ 0805/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective group I mGlu agonist; active enantiomer of 3,5-DHPG (Cat. No. 0342)

(S)-3,5-DHPG is a selective group I mGlu receptor agonist. (S)-3,5-DHPG regulates the expression of microRNAs in mouse cerebral cortex in vivo. (S)-3,5-DHPG induces a profound depression of synaptic dopamine release in mouse cortico-striatal cells in vitro, and anxiety-like behavior in vivo in mice.

Racemate also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162870-29-3
Molecular Formula C8H9NO4

Specifications

Quantity 5 mg
Formula Weight 183.16
Purity >98%
Molecular Weight (g/mol) 183.16
Chemical Name or Material (S)-3, 5-DHPG
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.