missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Hexanol, 98%

Product Code. 11398405
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11398405

Brand: Thermo Scientific Alfa Aesar L10401.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 52019-78-0 | C6H14O | 102.18 g/mol

S)-(+)-2-Hexanol is the S enantiomer used in the preparation of key intermediates for model studies in the total synthesis of antivirally active glycolipid cycloviracin B1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
S)-(+)-2-Hexanol is the S enantiomer used in the preparation of key intermediates for model studies in the total synthesis of antivirally active glycolipid cycloviracin B1.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52019-78-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00065955
InChI Key QNVRIHYSUZMSGM-UHFFFAOYNA-N
Synonym s-+-2-hexanol, s-2-hexanol, 2s-hexan-2-ol, 2-hexanol #, pubchem6732, 2s-2-hexanol, s-hexan-2-ol, s +-2-hexanol
PubChem CID 638097
IUPAC Name (2S)-hexan-2-ol
SMILES CCCCC(C)O

Specifications

Density 0.818
Boiling Point 137°C to 138°C
Flash Point 45°C (113°F)
Refractive Index 1.414
Quantity 1 g
UN Number UN2282
Beilstein 1718997
Solubility Information Slightly soluble in water.
Optical Rotation +10.5° (Neat)
Formula Weight 102.18
Percent Purity 98%
Chemical Name or Material (S)-(+)-2-Hexanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.