Learn More
S-2-Benzothiazolyl (Z)-2-(2-aminothiazole-4-yl)-2-methoxy-carbonylmethioacetate, TRC
Product Code. 30336002
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336002 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30336002 Supplier Toronto Research Chemicals Supplier No. TRCB41120310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material S-2-Benzothiazolyl (Z)-2-(2-aminothiazole-4-yl)-2-methoxy-carbonylmethioacetate
CAS 246035-38-1
Molecular Formula C15H12N4O4S3
Quantity 10 mg
Synonym Cefixime Impurity 2
InChI Formula InChI=1S/C15H12N4O4S3/c1-22-11(20)6-23-19-12(9-7-24-14(16)17-9)13(21)26-15-18-8-4-2-3-5-10(8)25-15/h2-5,7H,6H2,1H3,(H2,16,17)/b19-12-
SMILES COC(=O)CO/N=C(/C1=CSC(=N1)N)\C(=O)SC2=NC3=CC=CC=C3S2
IUPAC Name methyl 2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-(1,3-benzothiazol-2-ylsulfanyl)-2-oxoethylidene]amino]oxyacetate
Molecular Weight (g/mol) 408.47
Formula Weight 408.0021