Learn More
S-2-Benzothiazolyl-2-amino-a-(methoxyimino)-4-thiazolethiolacetate, TRC
Specifications
Specifications
| Chemical Name or Material | S-Benzothiazol-2-yl-(2Z)-(2-aminothiazol-4-yl)(methoxyimino)thioacetate |
| CAS | 80756-85-0 |
| Molecular Formula | C13 H10 N4 O2 S3 |
| Quantity | 10 g |
| Synonym | 4-Thiazoleethanethioic acid, 2-amino-α-(methoxyimino)-, S-2-benzothiazolyl ester, (αZ)-, 4-Thiazoleethanethioic acid, 2-amino-α-(methoxyimino)-, S-2-benzothiazolyl ester, (Z)-, (Z)-2-[Methoxyimino]-2-(2-aminothiazol-4-yl)acetic acid S-2-benzothiazole ester, (αZ)-2-Amino-α-(methoxyimino)-4-thiazoleethanethioic acid S-2-benzothiazolyl ester, 2-Amino-α-(methoxyimino)-4-thiazoleethanethioic acid S-2-benzothiazolyl ester, S-(Benzothiazol-2-yl) (Z)-(2-aminothiazol-4-yl)(methoxyimino)thioacetate, S-(Benzothiazol-2-yl) (Z)-(2-aminothiazol-4-yl)(methoxyimino)thioacetate, S-(Benzothiazol-2-yl) 2-amino-α-(methoxyimino)thiazole-4-thioacetate, S-1,3-Benzothiazol-2-yl (2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)ethanethioate, S-Benzothiazol-2-yl 2-(2-aminothiazol-4-yl)-2-(syn-methoxyimino)thioacetate |
| InChI Formula | InChI=1S/C13H10N4O2S3/c1-19-17-10(8-6-20-12(14)15-8)11(18)22-13-16-7-4-2-3-5-9(7)21-13/h2-6H,1H3,(H2,14,15)/b17-10- |
| SMILES | CO\N=C(/C(=O)Sc1nc2ccccc2s1)\c3csc(N)n3 |
| IUPAC Name | S-(1,3-benzothiazol-2-yl) (2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoethanethioate |
| Molecular Weight (g/mol) | 350.44 |
| Formula Weight | 349.9966 |