Learn More
(S)-2-Amino-6-(2,2,2-trifluoroacetamido)hexanoic acid, 97%, BLD Pharm™
Specifica
Specifica
| CAS | 10009-20-8 |
| Molecular Formula | C8H13F3N2O3 |
| MDL Number | MFCD00037223 |
| Quantity | 25 g |
| SMILES | N[C@@H](CCCCNC(C(F)(F)F)=O)C(O)=O |
| Molecular Weight (g/mol) | 242.20 |
| Percent Purity | 97% |
| Chemical Name or Material | (S)-2-Amino-6-(2,2,2-trifluoroacetamido)hexanoic acid |