missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(1-Boc-2-pyrrolidinyl)acetic acid, 95%, Thermo Scientific™

Product Code. p-7034489
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428727 250 mg 250mg
15418727 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428727 Supplier Thermo Scientific Alfa Aesar Supplier No. H52175.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56502-01-3
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD01862939
InChI Key GDWKIRLZWQQMIE-QMMMGPOBSA-N
Synonym boc-l-beta-homoproline, s-2-1-tert-butoxycarbonyl pyrrolidin-2-yl acetic acid, boc-beta-hopro-oh, 2s-1-tert-butoxycarbonyl pyrrolidin-2-yl acetic acid, boc-beta3-homopro-oh, s-2-1-boc-2-pyrrolidinyl acetic acid, s-1-boc-2-pyrrolidineacetic acid, 2-pyrrolidineacetic acid, 1-1,1-dimethylethoxy carbonyl-, 2s, 2-2s-1-tert-butoxycarbonylpyrrolidin-2-yl acetic acid, s-2-carboxymethyl-pyrrolidine-1-carboxylic acid tert-butyl ester
PubChem CID 688605
IUPAC Name 2-[(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidin-2-yl]acetic acid
SMILES CC(C)(C)OC(=O)N1CCCC1CC(=O)O

Specifications

Quantity 250 mg
Beilstein 478306
Formula Weight 229.28
Percent Purity 95%
Chemical Name or Material (S)-2-(1-Boc-2-pyrrolidinyl)acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.