Learn More
(S)-1-(3-Chlorophenyl)-2-hydroxy-1-propanone, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-1-(3-Chlorophenyl)-2-hydroxy-1-propanone |
| CAS | 287477-53-6 |
| Molecular Formula | C9H9ClO2 |
| Quantity | 10 mg |
| Synonym | Bupropion (S)-Hydroxy Propanone Impurity |
| InChI Formula | InChI=1S/C9H9ClO2/c1-6(11)9(12)7-3-2-4-8(10)5-7/h2-6,11H,1H3/t6-/m0/s1 |
| SMILES | CC(O)C(=O)c1cccc(Cl)c1 |
| IUPAC Name | (2S)-1-(3-chlorophenyl)-2-hydroxypropan-1-one |
| Molecular Weight (g/mol) | 184.62 |
| Formula Weight | 184.0291 |