Learn More
RWDD4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557690
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TYTLFEYAKD NKEQFMENHN PINSATSISN IISIETPNTA PSSKKKDKKE QLSKAQKRKL ADKTDHKGEL PRGWNWVDVV KHLSKTGSKD D Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.
RWDD4 Polyclonal antibody specifically detects RWDD4 in Human samples. It is validated for Immunohistochemistry.Specifications
RWDD4 | |
Polyclonal | |
Unconjugated | |
RWDD4 | |
BC016198; FAM28A; family with sequence similarity 28, member A; Gm1942; Protein FAM28A; RWD domain containing 4; RWD domain containing 4A; RWD domain-containing protein 4; RWD domain-containing protein 4A; Rwdd4; Rwdd4a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
201965 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NW29 | |
RWDD4 | |
Recombinant protein corresponding to Human RWDD4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only