missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RVG29-Cys

Product Code. 30564800
Change view
Click to view available options
:
5 mg
Product Code.
30564800 5 mg
1 options
This item is not returnable. View return policy

Product Code. 30564800

Brand: Tocris Bioscience™ 8903/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nicotinic acetylcholine receptor targeting peptide for neuronal delivery

RVG29-Cys is a peptide derived from rabies virus glycoprotein (RVG) able to specifically bind on the nicotinic acetylcholine receptors (nAchR) present on neuronal cells. RVG29-Cys can be used to functionalize LNPs via thiol-maleimide click chemistry for targeting nAchR present on blood-brain barrier (BBB). mRNA LNPs functionalized with RVG29-Cys shows improved transfection efficiency in cultured brain endothelial and neuronal cells, and in vivo, after systemic administration in mice.
TRUSTED_SUSTAINABILITY

Specifications

Purity 95%
CAS 1404289-25-3
Content And Storage Store at -20°C
Molecular Weight (g/mol) Observed: 3369.79
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.