Learn More
IKA™ RV 10.84 Evaporation Flask
Product Code. 11845922 Shop All IKA Products
Change view
Click to view available options
Volume (Metric):
1000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11845922 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11845922 Supplier IKA™ Supplier No. 0003740500
  • ONLINE EXCLUSIVE PRICE
    96.45€
    93.50€ / Pack of 1
    Save 2.95€
    3% Off
Estimated Shipment: 02-12-2026
High-quality borosilicate glass flask designed for evaporation processes.
  • High-quality borosilicate glass construction
  • Suitable for evaporation processes
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass
Type Evaporating Flask
Standard Taper Joint Number Center NS 29/32
Volume (Metric) 1000 mL