Learn More
Rucaparib, TRC
Product Code. 30133183
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133183 10 mg 10mg
30132451 50 mg 50mg
30006972 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133183 Supplier Toronto Research Chemicals Supplier No. TRCR70158010MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 283173-50-2
Molecular Formula C19H18FN3O
Molecular Weight (g/mol) 323.36
Synonym 8-Fluoro-1,3,4,5-tetrahydro-2-[4-[(methylamino)methyl]phenyl]-6H-pyrrolo[4,3,2-ef][2]benzazepin-6-one, 8-Fluoro-2-[4-[(methylamino)methyl]phenyl]-1,3,4,5-tetrahydro-6H-azepino[5,4,3-cd]indol-6-one, Rubraca, 8-Fluoro-1,3,4,5-tetrahydro-2-[4-[(methylamino)methyl]phenyl]-6H-azepino[5,4,3-cd]indol-6-one;
SMILES CNCc1ccc(cc1)c2[nH]c3cc(F)cc4C(=O)NCCc2c34
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C19H18FN3O/c1-21-10-11-2-4-12(5-3-11)18-14-6-7-22-19(24)15-8-13(20)9-16(23-18)17(14)15/h2-5,8-9,21,23H,6-7,10H2,1H3,(H,22,24)
Formula Weight 323.1434
Chemical Name or Material Rucaparib