Learn More
Fisherbrand™ Cork Stoppers
Product Code. 12376467 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Bottom:
6 mm
8 mm
9 mm
12 mm
13 mm
14 mm
17 mm
19 mm
21 mm
22 mm
25.5 mm
28.5 mm
30 mm
32 mm
35 mm
38 mm
41.5 mm
44 mm
47.5 mm
50 mm
Diameter (Metric) Top:
9.5 mm
11 mm
12 mm
15 mm
16 mm
17 mm
20.5 mm
22 mm
24 mm
25.5 mm
28.5 mm
32 mm
33 mm
35 mm
38 mm
41.5 mm
44 mm
47.5 mm
50 mm
57 mm
Unit Size:
Pack of 10
Pack of 20
Product Code. 12376467
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use in education, homebrewing and arts and crafts industries.
Available in different sizes
Specifications
Length (Metric) 32 mm
Quantity 10/Pk.
Diameter (Metric) Bottom 35 mm
Diameter (Metric) Top 38 mm
Material Cork
Product Type Cork Stopper