Learn More
Thermo Scientific™ Accessories for Digital Vortex Mixers
Product Code. 16574802 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
12 x 15 mm dia. Tube Holder
13 x 12 mm dia. Tube Holder
19 x 10 mm dia.Tube Holder
4 x 26 mm dia.Tube Holder
7 x 20 mm dia. Tube Holder
Flat Bottom Vessel Platform
Microplate Tray
Rubber Platform
Dimensions (L x W):
136.1 x 97.3 mm
82 x 16.5 mm
87.5 x 10 mm
87.5 x 40 mm
87.5 x 55 mm
Material:
Foam, plastic
Plastic
Rubber
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Dimensions (L x W) Material Unit Size
16574802 Microplate Tray 136.1 x 97.3 mm Plastic Each
16554802 Rubber Platform 82 x 16.5 mm Plastic Each
16564802 Flat Bottom Vessel Platform 87.5 x 10 mm Rubber Each
16584802 19 x 10 mm dia.Tube Holder 87.5 x 40 mm Foam, plastic Each
16594802 13 x 12 mm dia. Tube Holder 87.5 x 40 mm Foam, plastic Each
16504812 12 x 15 mm dia. Tube Holder 87.5 x 55 mm Foam, plastic Each
16514812 7 x 20 mm dia. Tube Holder 87.5 x 55 mm Foam, plastic Each
16524812 4 x 26 mm dia.Tube Holder 87.5 x 55 mm Foam, plastic Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16574802 Leverandør Thermo Scientific™ Leverandørnr. 88882122
  • ONLINE EXCLUSIVE PRICE
    21.65€
    20.10€ / Pack of 1
    Save 1.55€
    7% Off
In Stock
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
Get assorted accessories for Digital Vortex Mixers
Assorted accessories for Digital Vortex Mixers
Serviceplan
Standard Thermo warranty guideline, provide repair service and free spare part within warranty
garanti
2 years
TRUSTED_SUSTAINABILITY
Tekniske data
Product Type Microplate Tray
Material Plastic
Dimensions (L x W) 136.1 x 97.3 mm
Color Black
Description Microplate Tray
For Use With (Equipment) Digital Vortex mixer
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.