Learn More
Fisherbrand™ Rubber Pipette Bulbs for Bulb or Cylindrical Pipettes
Excellent for all media that do not attack rubber. Fisherbrand™ Pipette Bulbs come with three glassball valves, adjustable by finger pressure.
Brand:  Fisherbrand™ 4705541
Product Code. 11582672
  • ONLINE EXCLUSIVE PRICE
    35.25€
    5.15€ / Each
    Save 30.10€
    85% Off
Only null left
Estimated Shipment: 19-01-2026
Description
  • Available in a range of colors
  • With ca. 60mL volume suitable for pipettes from 2 to 50mL
  • Pipette media must not enter pipette filler; it may cause valve malfunction