Learn More
Fisherbrand™ Rubber Pipette Bulbs for Bulb or Cylindrical Pipettes
Excellent for all media that do not attack rubber. Fisherbrand™ Pipette Bulbs come with three glassball valves, adjustable by finger pressure.
Brand:  Fisherbrand™ 4705511
3.11 EUR valid until 2026-01-16
BEST PRICE on Promo! Use promo code "24103" to get your promotional price.
Product Code. 11552672
  • ONLINE EXCLUSIVE PRICE
    29.85€
    5.15€ / Each
    Save 24.70€
    82% Off
View all promotions
Only null left
In Stock
Edge
Add to basket
Description
Description
  • Available in a range of colors
  • With ca. 60mL volume suitable for pipettes from 2 to 50mL
  • Pipette media must not enter pipette filler; it may cause valve malfunction