Learn More
Abeba™ Rubber 9200 Shoes
Product Code. 15693138
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15693138 40 1 pair
15653138 36 1 pair
15663138 37 1 pair
15673138 38 1 pair
15683138 39 1 pair
15603148 41 1 pair
15613148 42 1 pair
15623148 43 1 pair
15633148 44 1 pair
15643148 45 1 pair
15653148 46 1 pair
15663148 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15693138 Supplier Abeba™ Supplier No. 9200/40
Only null left
Add to basket
Add to basket
Acid and base resistant. Rubber sole provides high slip resistance.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • White
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color White
Material Leather
Size 40
Sole Features Slip-resistant
Sole Material Rubber
Style 9200