missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RSeT™ Medium (2-Component)

Product Code. 17148311
Encompass
Click to view available options
Quantity:
500 mL Kit
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 17148311

Brand: STEMCELL Technologies 05978

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Defined medium for naïve-like human pluripotent stem cells

  • RSeT™ Medium is a defined cell culture medium used for the reversion of primed human pluripotent stem cells (hPSCs) to a naïve-like state and maintenance of naïve-like hPSCs under feeder-dependent and hypoxic conditions
  • RSeT™ Medium contains pre-screened quality components and does not contain bFGF or TGFβ
  • It is compatible with human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells
  • hPSCs cultured in RSeT™ Medium exhibit features of a naïve-like state such as tightly packed, domed colonies with refractive edges
  • Key transcripts associated with naïve-like hPSCs such as KLF2, KLF4, and TFCP2L1 show increased expression in hPSCs cultured in RSeT™ Medium
  • RSeT™ hPSCs can be converted back to a primed state by culture in mTeSR™1 and can then be differentiated with STEMdiff™ products such as STEMdiff™ Definitive Endoderm Kit (Catalog #05110), STEMdiff™ SMADi Neural Induction Kit (Catalog #08581), and STEMdiff™ Mesoderm Induction Medium (Catalog #05220)

Specifications

Quantity 500 mL Kit
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.