Learn More
R&D Systems™ (RS)-(Tetrazol-5-yl)glycine
Product Code. 18095332
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18095332 50 mg 50mg
18054662 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18095332 Supplier R&D Systems™ Supplier No. 0312/50
 Restricted Item
Estimated Shipment: 11-11-2026
A Tocris product, part of R&D Systems. Highly potent NMDA agonist
(RS)-(Tetrazol-5-yl)glycine is a potent NMDA receptor agonist, approximately 20 times more active than NMDA.
TRUSTED_SUSTAINABILITY
Specifications
CAS 138199-51-6
Assay Percent Range >98%
Quantity 50 mg
Solubility Soluble to 50 mM in water
Product Line Tocris
Target NMDA Receptor Agonists
Molecular Formula C3H5N5O2
Formula Weight Observed MW: 143.1
Purity 98%